Learn More
ORM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 164391AP
Catalog No. 50-173-0943
Add to cart
Description
Description
This antibody recognizes a band about 44 kDa in human plasma which may be due to the glycosylation of ORM1 or the dimer formation of the protein.
ORM1 is an acute phase plasma protein synthesized by the liver. The protein is believed to regulate the interaction between blood cells and endothelial cells, and together with haptoglobin and C reactive protein, also regulates the extravasation of the cells during infection and inflammation. ORM1 is a major acute phase reactant, both in human and in rat. In human plasma, ORM1 (44 kDa) is found at levels of 0.5-1.4 mg/mL. Its concentration in blood plasma is elevated during acute inflammation, such as trauma, maligcies, myocardial infarction, rheumatoid arthritis, bacterial infections, after major surgery, in cases of chronic pain etc., and can be used for the diagnosis of inflammatory conditions.
Specifications
Specifications
ORM1
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P02763, Q60590
AGP 1, AGP A, AGP1, Alpha 1 acid glycoprotein 1, OMD 1, ORM, ORM1, orosomucoid 1
Rabbit
IgG
150 μL
-20°C
Primary
18405, 5004
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ORM1
Liquid
Orm1
ORM1 Fusion Protein Ag9758
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse