Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ornithine Decarboxylase Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: rODC1/487, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314027UV
This item is not returnable.
View return policy
Description
Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Ornithine Decarboxylase | |
Monoclonal | |
DyLight 350 | |
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
Recombinant full-length human Ornithine Decarboxylase protein (Uniprot: P11926) | |
0.1 mL | |
Cell Cycle and Replication | |
4953 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence | |
rODC1/487 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title