Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ornithine Decarboxylase Mouse, Clone: SPM565, Novus Biologicals™
Mouse Monoclonal Antibody
$229.00 - $459.00
Specifications
Antigen | Ornithine Decarboxylase |
---|---|
Clone | SPM565 |
Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP232888A
|
Novus Biologicals
NBP2328880.1MG |
0.1 mg |
Each for $459.00
|
|
NBP232888F
|
Novus Biologicals
NBP2328880.02MG |
0.02 mg |
Each for $229.00
|
|
NBP232888B
|
Novus Biologicals
NBP2328880.2MG |
0.2 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Ornithine Decarboxylase Monoclonal specifically detects Ornithine Decarboxylase in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Ornithine Decarboxylase | |
Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
P11926 | |
4953 | |
Recombinant human ODC-1 protein | |
Primary | |
Store at 4C. | |
53 kDa |
SPM565 | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
ODC1 | |
IgG1 κ | |
Protein A purified | |
Recognizes a 53kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title