Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ornithine Decarboxylase Rabbit anti-Human, Mouse, Rat, Clone: ODC1/2878R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Ornithine Decarboxylase
Clone ODC1/2878R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122414
View Documents
Novus Biologicals
NBP30747020UG
20 μg
Each for $159.82
Add to cart
 
NB122413
View Documents
Novus Biologicals
NBP30747010UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

Ornithine Decarboxylase
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Purified
RUO
Human, Mouse, Rat
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1
Recombinant full-length human Ornithine Decarboxylase protein
Monoclonal
Protein A or G purified
ODC1/2878R
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml
Unconjugated
Rabbit
Cell Cycle and Replication
10 mM PBS with 0.05% BSA
4953
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit