Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orotic acid monohydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
50887-69-9
C5H4N2O4·H2O
174.11
MFCD00149398
YXUZGLGRBBHYFZ-UHFFFAOYSA-N
orotic acid monohydrate,orotic acid hydrate,2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate,lactinium,oroticum acidum,2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid,orotc acd hydrate?,uracil-6-carboxylic acid monohydrate,uracil-4-carboxylic acid hydrate
16218504
2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
C1=C(NC(=O)NC1=O)C(=O)O.O
50887-69-9
C5H4N2O4·H2O
174.11
MFCD00149398
YXUZGLGRBBHYFZ-UHFFFAOYSA-N
orotic acid monohydrate,orotic acid hydrate,2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate,lactinium,oroticum acidum,2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid,orotc acd hydrate?,uracil-6-carboxylic acid monohydrate,uracil-4-carboxylic acid hydrate
16218504
2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
C1=C(NC(=O)NC1=O)C(=O)O.O
Melting Point 334°C
Quantity 100g
Color White
Loss on Drying 12.0% max. (1 g, 130°C, 3 hrs)
Packaging Plastic bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Beilstein 25, 253
Merck Index 15, 6972
Solubility Information Solubility in water: 1.3g/L (20 c). Other solubilities: insoluble in chloroform
Formula Weight 174.11
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Orotic acid monohydrate
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.