Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259119

Catalog No. NBP259119


Add to cart

Description

Description

ORP1 Polyclonal antibody specifically detects ORP1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ORP1
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
FLJ10217, ORP1oxysterol-binding protein-related protein 1, ORP-1oxysterol-binding protein-related protein 1 variant 1, OSBP8, OSBPL1, OSBPL1B, OSBP-related protein 1, oxysterol binding protein-like 1A, oxysterol binding protein-like 1B, oxysterol-binding protein-related protein 1 variant 2
Rabbit
IgG
100 ul
Lipid and Metabolism
Primary
114876
Immunoprecipitation, Western Blot
Western Blot 1:1000, Immunoprecipitation
Affinity Purified
OSBPL1A
Maps to a region between 1 and 50
Immunogen affinity purified
RUO
Store at 4C.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only