Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORP150/HSP12A Antibody (6G7-2H5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259346H
Description
ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot.Specifications
ORP150/HSP12A | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
HYOU1 | |
Raised against a synthetic peptide of human GRP170 | |
100 μL | |
Primary | |
Detects 170kDa. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot | |
6G7-2H5 | |
Western Blot | |
150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
Mouse | |
Protein G purified | |
Hypoxia | |
10525 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction