Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORP150/HSP12A Antibody (6E3-2C3), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259345R
Description
ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ORP150/HSP12A | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HYOU1 | |
Recombinant Full length GRP170 Protein | |
100 μL | |
Primary | |
Detects 170 kDa. | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Immunocytochemistry, Immunofluorescence | |
6E3-2C3 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
Mouse | |
Protein G purified | |
Hypoxia | |
10525 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction