Learn More
ORP150/HSP12A Mouse anti-Human, Mouse, Rat, FITC, Clone: 6E3-2C3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259345F
Catalog No. NBP259345F
Add to cart
Description
Description
ORP150/HSP12A Monoclonal antibody specifically detects ORP150/HSP12A in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ORP150/HSP12A
6E3-2C3
Immunocytochemistry/Immunofluorescence
Purified
HYOU1
Full length GRP170 protein
Protein G purified
Hypoxia
Primary
10525
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD)
Mouse
IgG2b
100 ul
Store at 4C in the dark.
Monoclonal
Detects 170kDa.