Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORP150/HSP12A Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299781AF647

Catalog No. NB062868


Add to cart

Description

Description

ORP150/HSP12A Polyclonal antibody specifically detects ORP150/HSP12A in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

ORP150/HSP12A
Alexa Fluor 647
50mM Sodium Borate
150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD)
Produced in rabbits immunized with purified, recombinant ORP150/HSP12A (NP_001124463.1; Met 695-Leu 994).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Hypoxia
Primary
10525
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit