Ortho™ Anti-D Sera and Rh-hr Typing Reagents: Bioclones
Manufacturer: Ortho-Clinical Diagnostics 6901939
 View more versions of this product
Catalog No. 23-287-452
  Check Availability
Add to Cart
Alert
Contains sodium azide. Sodium azide may react with lead and copper plumbing to form highly explosive metal azide. On disposal, flush with a large volume of water to prevent azide buildup.
Related Products
Product Content Feedback