Ortho™ Bovine Albumin for Blood Typing Reactions
Enhances antibody reactions
Manufacturer: Ortho-Clinical Diagnostics 718300
 View more versions of this product
Catalog No. 23-287-484
  Check Availability
Add to Cart
    Specifications
    Product Type Polymerized Bovine Albumin Solution
    For Use With (Application) For Blood typing reactions, enhance Antigen/antibody interactions, increase in speed and sensitivity of detection
    Quantity 100 x 10mL
  • Description & Specifications
Related Products
Product Content Feedback