Ortho-Clinical Diagnostics™ VITROS™ MicroSlides

For use with the Ortho VITROS™ 250 System

Manufacturer: Ortho-Clinical Diagnostics 6802469

 View more versions of this product

Catalog No. 23-610-055

 N/A
N/A

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 


    Specifications

    Product Type Direct HDL Cholesterol-dHDL
    For Use With (Application) In-vitro diagnostic use
    For Use With (Equipment) Vitros 5,1 FS Chemistry System
    Formulation Not Applicable
    Quantity 5/Pk., 90 Slides
    Sample Size Not Available
    Sample Type Not Available

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit