Learn More
Ortho-Clinical Diagnostics™ VITROS™ MicroSlides Available on GSA/VA Contract for Federal Government customers only.
For use with the Ortho VITROS™ 250 System
Manufacturer:  Ortho-Clinical Diagnostics 1336544
 View more versions of this product
Catalog No. 23-046-161
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This product is regulated by the federal government, and may not be ordered online.
Specifications Description & Specifications
Specifications
In-vitro diagnostic use
Lipase (Candida rugosa, E.C.3.1.1.3) 0.15U, Peroxidase (horseradish root, E.C.1.11.1.7) 0.52U, Glycerol Kinase (Cellulomonas sp., E.C.2.7.1.30) 0.35U, L-alpha-Glycerophosphate Oxidase (Pediococcus sp., E.C.1.1.3.-) 0.19 U, Triton X-100 0.62mg, 2-(3,5-Dimethoxy-4-Hydroxyphenyl)-4,5-bis(4-Dimethylaminophenyl) Imidazole (leuco dye) 0.04mg and Adenosine Triphosphate 0.14mg, Pigment, Binders, Buffer, Surfactants, Stabilizers, scavenger, Enzyme cofactors, dye solubilizer and cross-linking agent
5.5μL
Triglyceride dry slide
Vitros™ 250 system
5 x 60 Pack
Heparin