Learn More
Ortho-Clinical Diagnostics™ VITROS™ MicroSlides
For use with the Ortho VITROS™ 250 System
Supplier:  Ortho-Clinical Diagnostics™ 1314343
 View more versions of this product
Catalog No. 23-046360
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
Description
Description
Specifications
Specifications
Heparin, Plasma, Serum
Salicylate Hydroxylase (Pseudomonas sp., E.C.1.14.13.1) 0.07U, Tyrosinase (mushroom, E.C.1.14.18.1) 0.12U, 3-Methyl-2-Benzothiazolinone Hydrazone Hydrochloride (MBTH) 40μg and Nicotinamide Adenine Dinucleotide 0.1mg, reduced, Pigment, Bi
5 x 18 Pack
Salicylate Dry Slide
In-vitro diagnostic use
10 μL