Learn More
Ortho-Clinical Diagnostics™ VITROS™ Thyroid-stimulating hormone (TSH)
Supplier:  Ortho-Clinical Diagnostics™ 1912997
Catalog No. 23-057-285
Due to product restrictions, please to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Thyroid Stimulating Hormone (TSH)
Vitros™ TSH Reagent
100 Pack
37 min.
Well
Coated wells 100, conjugate reagent 6.2mL, Biotinylated Antibody reagent 9.1mL
80 μL