Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics VITROS 250/350 Controls
Controls for Ortho Clinical VITROS 250/350
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity & Availability | ||||
23-046189
|
Ortho-Clinical Diagnostics
8748220 |
Isoenzyme Performance Verifier II | 6 ST | N/A | N/A | ||||
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. This product is regulated by the federal government, and may not be ordered online. |
|||||||||
23-046184
|
Ortho-Clinical Diagnostics
8067324 |
Performance Verifier I | 12 ST | N/A | N/A | ||||
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. This product is regulated by the federal government, and may not be ordered online. |
|||||||||
23-046185
|
Ortho-Clinical Diagnostics
8231474 |
Performance Verifier II | 12 ST | N/A | N/A | ||||
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. This product is regulated by the federal government, and may not be ordered online. |
|||||||||
23-046186
|
Ortho-Clinical Diagnostics
8466492 |
Liquid Performance Verifier I | 6 ST | N/A | N/A | ||||
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. This product is regulated by the federal government, and may not be ordered online. |
|||||||||
23-046187
|
Ortho-Clinical Diagnostics
1384007 |
Liquid Performance Verifier II | 6 ST | N/A | N/A | ||||
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. This product is regulated by the federal government, and may not be ordered online. |
|||||||||
23-046188
|
Ortho-Clinical Diagnostics
8721508 |
Isoenzyme Performance Verifier I | 6 ST | N/A | N/A | ||||
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. This product is regulated by the federal government, and may not be ordered online. |
|||||||||
23-046190
|
Ortho-Clinical Diagnostics
8962540 |
TDM Performance Verifier I | 6 ST | N/A | N/A | ||||
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. This product is regulated by the federal government, and may not be ordered online. |
|||||||||
23-046191
|
Ortho-Clinical Diagnostics
1792357 |
TDM Performance Verifier II | 6 ST | N/A | N/A | ||||
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. This product is regulated by the federal government, and may not be ordered online. |
|||||||||
23-046192
|
Ortho-Clinical Diagnostics
8182172 |
TDM Performance Verifier III | 6 ST | N/A | N/A | ||||
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. This product is regulated by the federal government, and may not be ordered online. |
|||||||||
Description
For use with Ortho Clinical VITROS 250 systems, Cat. Nos. 22-046-351, -353
Specifications
Ortho clinical VITROS™ 250 system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title