Learn More
Ortho Clinical VITROS™ 250/350 Dry Slides (A to L)
Manufacturer:  Ortho-Clinical Diagnostics 1515808
 View more versions of this product
Catalog No. 23-046181
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This product is regulated by the federal government, and may not be ordered online.
Specifications Description & Specifications
Specifications
In-vitro diagnostic use
Dry slide
Each
Iron/TIBC slide
Vitros™ 250 system
Ascorbic Acid 160μg and N-(4-(2,4-bis(1,1-dimethylpropyl) phenoxy)butyl)- 5-methoxy-6((2,3,6,7-tetrahydro-8-1H, 5Hbenzo-(ij)-quinolizin-9-yl)azo)- 3-pyridine sulfonamide (dye) 5μg, Binders, Buffer, Pigment, Surfactants,
Heparin