Ortho Clinical VITROS™ 250/350 Dry Slides (A to L)

Manufacturer: Ortho-Clinical Diagnostics 1037274

 View more versions of this product

Catalog No. 23-046363

 N/A
N/A

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 


    Specifications

    Product Type 18-Acetaminophen slide
    For Use With (Application) In-vitro diagnostic use
    For Use With (Equipment) Vitros™ 250 system
    Format Dry slide
    Formulation Aryl Acyl Amidase (Pseudomonas, E.C.3.5.1.13) 0.3U, Potassium Ferricyanide 0.18mg, 1-(3-Sulfopropyl-1,2,3,4-Tetrahydroquinoline) 0.1mg, Pigment, Binders, Buffer, Surfactants, scavenger and cross-linking agent
    Quantity 108 Pack
    Sample Type Serum, Plasma, Heparin

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit