Ortho Clinical VITROS™ 250/350 Dry Slides (A to L) Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Ortho-Clinical Diagnostics 8297749

 View more versions of this product

Catalog No. 23-046372

    N/A

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Sample Type Heparin
    Product Type 18-Lipase slide
    For Use With (Application) In-vitro diagnostic use
    For Use With (Equipment) VITROS 250 Dry Slides
    Format Dry slide
    Formulation Diacetinase (Bacillus subtilis, E.C.3.1.1-) 0.54U, Glycerol Kinase (E. coli or Cellulomonas sp, E.C.2.7.1.30) 0.32U, L-alpha-Glycerophosphate Oxidase (Aerococcus viridans, E.C.1.1.3.21) 0.39U, Peroxidase (horseradish root, E.C.1.11.1.7) 0.62U, Colipase (porcine pancreas) 5.9U, Adenosine Triphosphate 0.16mg, 1-Oleoyl-2,3-Diacetylglycerol 0.8mg and 2-(3,5-Dimethoxy- 4-Hydroxyphenyl)-4,5-bis(4-Dimethylaminophenyl) Imidazole (Leuco dye) 33μg, Pigment, Binders, Surfactants, Enzyme cofactors, Stabilizers, Buffer, dye solubilizer, scavenger and cross-linking agent
    Quantity Each

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit