Ortho Clinical VITROS™ 250/350 Dry Slides (A to L)

Manufacturer: Ortho-Clinical Diagnostics 8097990

 View more versions of this product

Catalog No. 23-044-667

    N/A

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Sample Type Heparin
    Product Type C-Reactive Protein
    For Use With (Application) In-vitro diagnostic use
    For Use With (Equipment) Vitros™ 250/350, 500, 750, 950 dry slides
    Formulation Immobilized Phosphorylcholine 0.07mg, mouse Anti-CRP antibody labeled with horseradish Peroxidase 0.0006U, Calcium Chloride 0.08mg, 2-(3,5-dimethoxy-4-hydroxyphenyl)-4,5-bis (4-dimethylaminophenyl) Imidazole (leuco dye) 0.04mg, Binders, pH 7 Buffer, Surfactants, cross-linking agent, Polymer beads, Proteins, Stabilizers and wash detection dye
    Quantity 5 cartridges of 18 slides
    Format Dry slide

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit