Learn More
Ortho Clinical VITROS™ 250/350 Dry Slides (A to L) Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Ortho-Clinical Diagnostics 8196057
 View more versions of this product
Catalog No. 23-046163
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
In-vitro diagnostic use
Dry slide
Each
Albumin slide
VITROS 250 Dry Slides
Bromocresol green dye 104μg, Polymer beads, Binders, Buffer pH 3.1 and Surfactants
Heparin