Learn More
Ortho Clinical VITROS™ 250/350 Dry Slides (A to L) Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Ortho-Clinical Diagnostics 1655281
 View more versions of this product
Catalog No. 23-046165
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This product is regulated by the federal government, and may not be ordered online.
Specifications Description & Specifications
Specifications
In-vitro diagnostic use
Dry slide
Each
Alanine Aminotransferase (SGPT) slide
VITROS 250 Dry Slides
Lactate Dehydrogenase (porcine muscle, E.C.1.1.1.27) 0.12U, Lalanine 0.86mg, Sodium a-ketoglutarate 54μg, Nicotinamide Adenine Dinucleotide, reduced 35μg and 11μg Sodium Pyridoxal-5-Phosphate, Pigment,
Heparin