Learn More
Ortho Clinical VITROS™ 250/350 Dry Slides (A to L) Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Ortho-Clinical Diagnostics 8433815
 View more versions of this product
Catalog No. 23-046166
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
In-vitro diagnostic use
Dry slide
Each
Aspartate Aminotransferase (SGOT)
VITROS 250 Dry Slides
Sodium Aspartate 0.27mg, Sodium a-Ketoglutarate 0.13mg, Sodium Pyridoxal-5-phosphate 11μg, Sodium Phosphate 42μg, 2- (3,5-dimethoxy- 4-hydroxyphenyl)-4,5-bis(4- dimethylaminophenyl) Imidazole (leuco dye) 30μg
Heparin