Learn More
Ortho Clinical VITROS™ 250/350 Dry Slides (A to L) Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Ortho-Clinical Diagnostics 8383051
 View more versions of this product
Catalog No. 23-046170
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This product is regulated by the federal government, and may not be ordered online.
Specifications Description & Specifications
Specifications
In-vitro diagnostic use
Dry slide
Each
BuBc slide
VITROS 250 Dry Slides
Caffeine 560μg, Sodium Benzoate 530μg and Mordant 180μg, Binders, Pigment, Buffer, Surfactants, Stabilizer and cross-linking agent
Heparin