Learn More
Ortho Clinical VITROS™ 250/350 Dry Slides (A to L)
Manufacturer:  Ortho-Clinical Diagnostics 8058232
 View more versions of this product
Catalog No. 23-046176
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This product is regulated by the federal government, and may not be ordered online.
Specifications Description & Specifications
Specifications
In-vitro diagnostic use
Dry slide
5 cartridges of 60 slides
Creatine Kinase MB slide
Vitros™ MicroSlides
L-alpha-glycerofosfat Oxidase (Aerococcus viridans, E.C.1.1.3.21) 039U, Peroxidase (peberrod, E.C.1.11.1.7) 1,4U, Glycerolkinase (E. coli eller Cellulomonas sp, E.C.2.7.1.30) 0.45U, Kreatinfosfat 0.17mg, N-acetylcystein 43μg, gede Antihu
Heparin