Ortho Clinical VITROS™ 250/350 Dry Slides (A to L) Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Ortho-Clinical Diagnostics 8046872

 View more versions of this product

Catalog No. 23-046361

 N/A
N/A

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 


    Specifications

    Sample Type Fluoride Oxalate
    Product Type 18-Alcohol slide
    For Use With (Equipment) Vitros™ 250 system
    Formulation Alcohol Dehydrogenase (Yeast, E.C.1.1.1.1) 1.1U, Nicotinamide Adenine Dinucleotide 0.8mg, Tris(hydroxymethyl) Aminomethane 1mg, Pigment, Binders, Stabilizer, Surfactants, protein and cross-linking agent
    Quantity Each
    For Use With (Application) In-vitro diagnostic use
    Format Dry slide

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit