Ortho Clinical VITROS™ DT60 II Solutions Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Ortho-Clinical Diagnostics 1448042

 View more versions of this product

Catalog No. 23-038786

 N/A
N/A

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Storage Requirements Refrigerated
    For Use With (Application) Control
    For Use With (Equipment) VITROS™ DT60 II chemistry analyzer
    Quantity 12 ST
    Level 2,2
    Product Type Kodatrol DT Control II

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit