Learn More
Ortho-Clinical Diagnostics™ VITROS™ Anti-Human immuno-deficiency virus (A-HIV) 1+2
Supplier:  Ortho-Clinical Diagnostics™ 6801861
Catalog No. 23-057-254
Due to product restrictions, please to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
HIV-1/2 Antibodies
Vitros™ Anti-HIV 1+2 Reagent
100 Pack
48 min.
Well
Coated wells 100, assay reagent 6.2mL, conjugate reagent 13.3mL
80 μL