Learn More
Ortho-Clinical Diagnostics™ VITROS™ Follicle-stimulating hormone (FSH)
Supplier:  Ortho-Clinical Diagnostics™ 1931922
Catalog No. 23-057-268
Due to product restrictions, please to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Follicle stimulating hormone (FSH)
Vitros™ FSH Reagent
100 Pack
24 min.
Well
Coated wells 100, conjugate reagent 6.2mL, Biotinylated Antibody reagent 13.3mL
40 μL