Osborn Scientific™ BADD™ (BioWarfare Agent Detection Device) Detection Sets
Sample from hard surfaces or liquid to test for anthrax, botulinum toxin, ricin toxin, and training
 View all options for this product
Catalog No. 22-900-104
  Check Availability
Add to Cart
    Includes Test cassette, swab, dropper, microtube, biosimulant-contaminated label and instructions
    Product Type Training test set
    For Use With (Application) BioWarfare Agent Training Test Set;
  • Description & Specifications
Related Products
Product Content Feedback