Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OSBP Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518364
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Oxysterol binding protein is an intracellular protein that is believed to transport sterols from lysosomes to the nucleus where the sterol down-regulates the genes for the LDL receptor, HMG-CoA reductase, and HMG synthetase.
Specifications
OSBP | |
Polyclonal | |
Unconjugated | |
Osbp | |
1110018F06Rik; AW559088; HGNC:8503; Kiaa4220; mKIAA4220; OSBP; OSBP1; oxysterol binding protein; oxysterol-binding protein 1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5007 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P22059 | |
Osbp | |
Synthetic peptide sequence (KHQLEETKKEKRTRIP) corresponding to the internal amino acids of OSBP (aa 395-411). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction