Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSBPL11 Mouse anti-Human, Clone: OTI10F6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502067

Catalog No. 50-167-3153


Add to cart

Description

Description

This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.
Specifications

Specifications

OSBPL11
Monoclonal
0.59 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BXB4
ORP-11, ORP11, OSBP12, TCCCIA00292
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
114885
Immunocytochemistry, Immunofluorescence, Western Blot
OTI10F6
Unconjugated
OSBPL11
Liquid
OSBPL11
Full length human recombit protein of human OSBPL11 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit