Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSBPL11 Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: OTI6A6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273160C
Description
OSBPL11 Monoclonal specifically detects OSBPL11 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
OSBPL11 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
OSBPL11 | |
Full length recombinant protein of human OSBPL11 (NP_073613) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI6A6 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
FLJ13012, FLJ13164, ORP11OSBP-related protein 11, ORP-11TCCCIA00292, OSBP12, oxysterol binding protein-like 11, oxysterol-binding protein-related protein 11 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
114885.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title