Learn More
OSBPL11 Mouse anti-Canine, Human, Non-human primate, Clone: OTI6H9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525293
Catalog No. PIMA525293
Add to cart
Description
Description
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.
Specifications
Specifications
OSBPL11
Monoclonal
0.96 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BXB4
OSBPL11; ORP11; ORP-11; OSBP12; FLJ13012; FLJ13164; TCCCIA00292; oxysterol binding protein-like 11; OSBP-related protein 11; oxysterol-binding protein-related protein 11
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100856388, 114885, 716096
Immunocytochemistry, Immunofluorescence, Western Blot
OTI6H9
Unconjugated
OSBPL11
Liquid
OSBPL11
Full length human recombit protein of OSBPL11 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate