Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OSBPL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110064
Description
Immunogen sequence: SSSPSAQVIG LKNALSSALA QNTDLKERLR RIHAESLLLD SPAVAKSGDN LAEENSRDEN RALVHQLSNE SRLSITDSLS EFFDAQEVLL SPSSSENEIS DDDSYVSDIS DNLSLDNLSN DLDNERQTLG PVLDSGREAK SRRRTCLPAP C.
ZNF703 belongs to the Elbow/Noc family. It contains 1 C2H2-type zinc finger.ZNF703 may function as a transcriptional repressor.
Specifications
OSBPL3 | |
Polyclonal | |
Unconjugated | |
OSBPL3 | |
1200014M06Rik; 6720421I08Rik; A530055M08; KIAA0704; ORP3; ORP-3; OSBP3; Osbpl3; OSBP-related protein 3; oxysterol binding protein like 3; oxysterol binding protein-like 3; oxysterol-binding protein-related protein 3; RGD1564287 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26031, 362360, 71720 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.86 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9DBS9, Q9H4L5 | |
OSBPL3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 370-520 of human OSBPL3 (NP_056365.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction