Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OSCAR Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547171
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Recently, a novel member of the leukocyte receptor complex (LRC) encoded family has been identified that is expressed specifically in osteoclast (OC). This protein names OC-associated receptor or OSCAR, play critical roles in the regulation of both innate and adaptive immune responses and bone-specific regulator of OC differentiation. Osteoclasts are multinucleated cells that resorb bone and are essential for bone homeostasis. This protein family plays critical roles in the regulation of both innate and adaptive immune responses. Different from the other LRC members, OSCAR expression is detected specifically in preosteoclasts or mature osteoclasts. OSCAR may be an important bone-specific regulator of osteoclast differentiation. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
OSCAR | |
Polyclonal | |
Unconjugated | |
OSCAR | |
MGC33613; mOSCAR; mOSCAR-M1; mOSCAR-M2; mOSCAR-M3; ocl178; Oscar; osteoclast associated receptor; osteoclast associated receptor OSCAR-S1; osteoclast associated receptor OSCAR-S2; osteoclast associated, immunoglobulin-like receptor; osteoclast-associated immunoglobulin-like receptor; Osteoclast-associated receptor; osteoclast-associated receptor OSCAR; PIGR3; PIgR-3; poly-Ig receptor 3; polymeric immunoglobulin receptor 3 | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
232790 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8VBT3 | |
OSCAR | |
Mouse myeloma cell line NS0-derived recombinant mouse OSCAR Asp19-Asn228. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction