Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSGEP Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI9E3, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503002
Description
Osgep (O-sialoglycoprotein endopeptidase), also known as GCPL1, is a 335 amino acid protein that is a member of the peptidase M22 family. Osgep specifically cleaves the 31-Arg-l-Asp-32 bond in glycophorin A, but it does not cleave desialylated glycoproteins, unglycosylated proteins or glycoproteins that are only N-glycosylated. Though ubiquitously expressed at low levels, highest levels of Osgep are found in liver, skeletal muscle and kidney. OSGEPL1, also known as Qri7, is a 414 amino acid protein that belongs to the KAE1/YgjD family and exists as three alternatively spliced isoforms. In tRNAs that have codons beginning with adenine, OSGEPL1 is required for the formation of a threonylcarbamoyl group on adenosine.Specifications
OSGEP | |
Monoclonal | |
0.82 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8BWU5, Q9NPF4, Q9WVS2 | |
GAMOS3, GCPL1, KAE1, OSGEP1, PRSMG1, TCS3 | |
Mouse | |
IgG2a | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
100687555, 290028, 55644, 66246 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI9E3 | |
Unconjugated | |
OSGEP | |
Liquid | |
Osgep | |
Human recombit protein fragment corresponding to amino acids 1-335 of human OSGEP produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Canine, Human, Mouse, Rat |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title