Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSGEP Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI9E3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525567

Catalog No. PIMA525567


Add to cart

Description

Description

Osgep (O-sialoglycoprotein endopeptidase), also known as GCPL1, is a 335 amino acid protein that is a member of the peptidase M22 family. Osgep specifically cleaves the 31-Arg-l-Asp-32 bond in glycophorin A, but it does not cleave desialylated glycoproteins, unglycosylated proteins or glycoproteins that are only N-glycosylated. Though ubiquitously expressed at low levels, highest levels of Osgep are found in liver, skeletal muscle and kidney. OSGEPL1, also known as Qri7, is a 414 amino acid protein that belongs to the KAE1/YgjD family and exists as three alternatively spliced isoforms. In tRNAs that have codons beginning with adenine, OSGEPL1 is required for the formation of a threonylcarbamoyl group on adenosine.
Specifications

Specifications

OSGEP
Monoclonal
0.82 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NPF4, Q8BWU5, Q9WVS2
Probable tRNA N6-adenosine threonylcarbamoyltransferaseUniRule annotation; N6-L-threonylcarbamoyladenine synthase; t(6)A synthase; O-sialoglycoprotein endopeptidaseUniRule annotation; hOSGEP; t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEPUniRule annotation; tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPUniRule annotation; OSGEP; GCPL1
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100687555, 290028, 55644, 66246
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI9E3
Unconjugated
OSGEP
Liquid
OSGEP
Human recombit protein fragment corresponding to amino acids 1-335 of OSGEP produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit