Learn More
OSGIN2 Mouse anti-Human, Clone: 2C4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524577
Catalog No. PIMA524577
Add to cart
Description
Description
Peptide Sequence: SGLKKIFKLS AAVVLIGSHP NLSFLKDQGC YLGHKSSQPI TCKGNPVEID TYTYECIKEA NLFALGPLVG DNFVRFLKGG ALGVTRCLAT RQKKKHLFVE RGGGDGIA
May be involved in meiosis or the maturation of germ cells.
Specifications
Specifications
OSGIN2
Monoclonal
Unconjugated
OSGIN2
Liquid
OSGIN2
OSGIN2 (AAH31054, 398 a.a. ∽ 505 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA
2C4
PBS with no preservative; pH 7.4
Q9Y236
C8orf1, hT41, oxidative stress-induced growth inhibitor 2
Mouse
IgG2b, kappa
100 μg
-20° C
Primary
734