Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OSGIN2 Monoclonal Antibody (OTI7D12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527078
Description
OSGIN2 Monoclonal Antibody for Western Blot, IHC (P)
May be involved in meiosis or the maturation of germ cells.
Specifications
OSGIN2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y236 | |
OSGIN2 | |
Human recombinant protein fragment corresponding to amino acids 1-250 of OSGIN2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7D12 | |
Unconjugated | |
OSGIN2 | |
BC010311; C230027H09Rik; C8orf1; hT41; LOW QUALITY PROTEIN: oxidative stress-induced growth inhibitor 2; OSGIN2; oxidative stress induced growth inhibitor family member 2; oxidative stress-induced growth inhibitor 2; RGD1306323 | |
Mouse | |
Affinity Chromatography | |
RUO | |
734 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction