Learn More
OSGIN2 Mouse anti-Human, Clone: OTI7D12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808943
Catalog No. 50-166-9185
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May be involved in meiosis or the maturation of germ cells.
Specifications
Specifications
OSGIN2
Monoclonal
Unconjugated
OSGIN2
Lyophilized
OSGIN2
Human recombit protein fragment corresponding to amino acids 1-250 of human OSGIN2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI7D12
PBS with 8% trehalose and no preservative; pH 7.3
Q9Y236
C8orf1, hT41
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
734