Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OSGIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555887
Description
Immunogen sequence: SNFIKRNWEI RGYQRIADGS HVPFCLFAEN VALATGTLDS PAHLEIEGED FPFVFHSMPE FGAAINKGKL RGKVDPVLIV GSG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
May be involved in meiosis or the maturation of germ cells.
Specifications
OSGIN2 | |
Polyclonal | |
Unconjugated | |
OSGIN2 | |
BC010311; C230027H09Rik; C8orf1; hT41; LOW QUALITY PROTEIN: oxidative stress-induced growth inhibitor 2; OSGIN2; oxidative stress induced growth inhibitor family member 2; oxidative stress-induced growth inhibitor 2; RGD1306323 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
734 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y236 | |
OSGIN2 | |
Recombinant protein corresponding to Human OSGIN2. Recombinant protein control fragment (Product #RP-93871). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction