Learn More
Selleck Chemical LLCSupplier Diversity Partner OSI-930 25mg 728033-96-3  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S122025MG
OSI-930 is a potent inhibitor of Kit (c-Kit), KDR and CSF-1R with IC50 of 80 nM, 9 nM and 15 nM, respectively; also potent to Flt-1, c-Raf and Lck and low activity against PDGFRα/β, Flt-3 and Abl. Phase 1.
Catalog No. 50-690-7