OSM™ All-Natural Degreaser Available on GSA/VA Contract for Federal Government customers only.

Cuts through grease and grime in seconds

Manufacturer: ORGANIC SALES AND MARKETING INC 81008201228

 View more versions of this product

Catalog No. 15-336-511

  Check Availability
Add to Cart

    Specifications

    Size 1 gal.
    Certifications/Compliance 29 CFR1910.1200
    Color Clear Light Blue
    Format Liquid
    For Use With (Application) Ideal for steam cleaning engines and machinery or for general cleanups in and around facilities

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit