OSM™ All-Natural No-Rinse Floor Cleaner Available on GSA/VA Contract for Federal Government customers only.

Cleans gently, leaving a beautiful luster; safe for use around people and animals

Manufacturer: ORGANIC SALES AND MARKETING INC 81008201217

 View more versions of this product

Catalog No. 15-336-501

  Check Availability
Add to Cart

    Specifications

    Color Green
    Format Liquid
    For Use With (Application) For use on tile, laminate, Vinyl, Wood and painted surfaces, safe for cleaning cabinets, molding, paneling and doors
    Product Type All-natural No-rinse floor cleaner

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit