OSM™ Natural Concrete Cleaner

Uses powerful natural cleaning agents to remove oil, grease and grime from concrete, asphalt, and stone

Manufacturer: ORGANIC SALES AND MARKETING INC 81008201267

 View all options for this product

Catalog No. 15-336-506


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Color Clear
    For Use With (Application) Oil, Grease or food stains on garage floors, parking lots and sidewalks, Perfect for use in power washers
    Product Type Natural concrete cleaner
    Format Liquid
    For Use With (Equipment) Power washer

    Ideal to use on driveways, patios, floors, and sidewalks.

    • Powerful cleaning agents remove petroleum-based stains on garage floors, parking lots, and sidewalks
    • Can be used in power washers
    • Renewable plant-derived formula free of phospahtes, glycol ether, and phthalate
    • Noncaustic and noncorrosive
    • Will not harm people, animals, wildlife, or lawn and grounds
    • Environmentally preferable, ozone safe formula disposes down the drain
    • No harsh chemical fumes or vapor

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit