Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precision Systems Osmette™ XL Osmometer

Automatic digital osmometers measure osmolality in mOsm/kg H2O. Precision Systems Osmette XL Osmometer uses the freezing point depression method to provide a quick, accurate indication of total concentration. Instruments measure freezing points of solutions to 0.001°C, then directly translate them to measure of concentration.

Manufacturer:  Precision Systems 5007

Catalog No. 12-827-33


Add to cart

Description

Description

Easy to Use

  • Add sample tube to well, lower operating head to insert wire and thermistor temperature probe into the sample
  • Freezing point is detected by the probe, with accuracy of ±0.001°C
  • Digital display shows results directly in mOsm/kg H2O
  • Entire cycle takes between one-half and one minute per sample
Speedy and convenient
  • Fast cooldown, completely zero-stable meter
  • Reach operating temperature in <15 min. vs. 40 to 60 min. for other cryoscopic devices
  • Eliminate Dewar flasks, ice baths, calculations
  • Generate precise results in just 50 to 70 sec.; process 40 to 60 samples per hour
  • Minimal training; simple two-step calibration takes less than five minutes
  • All models hold calibration, eliminating daily calibration

Osmette XL, Model 5007

  • Microprocessor-controlled calibration
  • Includes built-in printer, keypad, statistical software and RS-232-C port
  • Small 2 oz. (59mL) bath allows faster results with better control
  •  Food and beverage industries, chemical manufacturing, pharmaceutical labs, research and quality control
  • Clinical applications include diagnosis of renal activity, disease electrolyte imbalance, hyper- and hyponatremia, measuring osmolality of body fluids

1 Reproducibility, 1 S.D. (one standard deviation).
2 With liquid bath.
Also available: Cryette™ Cryoscopes. Model 5006 (Cat. No. 12-672) determines added water in milk; Models 5008 and 5009 (Cat. Nos. 12-672-1 and 12-672-2) determine molecular weight of solutions such as petroleum fractions and core samples. For information, contact your Fisher Scientific Customer Service Representative.

Specifications

Specifications

50/60Hz
Thermoelectric cooler
Osmette™ Xl Osmometer
Digital
10 in.
8 in.
20cm
For 2mL sample: <1000mOsm/kg: ±1mOsm/kgH2O, >1000mOsm/kgH2O: ±0.2%. For 0.2mL sample: <500mOsm/kg: ±2mOsm/kgH2O, >500mOsm/kgH2O: ±0.5%
2 or 0.2mL
15 in.
8 x 15 x 10 in. (20 x 38 x 25cm)
115V
Each
5007
Food and beverage industries, chemical manufacturing, pharmaceutical labs, research and quality control, clinical applications include diagnosis of renal activity, disease electrolyte imbalance, hyper- and hyponatremia, measuring osmolality of body fluids
25cm
<1 min.
Automatic
0 to 3000mOsm/kg H2O
38cm
10 to 15 min.
115V 50/60Hz
SDS
Documents
Product Certifications
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit