Osmic Acid Anhydride (Crystalline/Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Safety seal, prescored neck, easy-to-snap-open ampules

Packaging Ampule
Quantity 0.5g

 View more versions of this product

Catalog No. A235-12

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 20816-12-0
    Physical Form Solid
    Molecular Formula O4Os
    Formula Weight 254.23
    MDL Number MFCD00011150
    Synonym Osmium Tetroxide
    Chemical Name or Material Osmic Acid Anhydride
    Grade Laboratory Grade

    Specifications

    Boiling Point 130°C
    Formula Weight 254.23
    Melting Point 39.5°C
    Quantity 0.5g
    DOT Information DOT Class 6.1, : Poison
    Viscosity [NULL]
    Physical Form Solid
    Color Off-white
    Density 4.9g/mL
    Vapor Pressure 7mmHg at 20°C
    Identification Pass Test
    Packaging Ampule
    Molecular Formula O4Os
    MDL Number MFCD00011150
    CAS 20816-12-0
    ChemAlert Storage Symbol White
    Synonym Osmium Tetroxide
    Chemical Name or Material Osmic Acid Anhydride
    Grade Laboratory Grade
     

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit