Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Osmium Tetroxide, Honeywell

Click to view available options
Quantity:
100mg
250mg
500mg
1g
20816-12-0
O4Os
254.226
MFCD00011150
VUVGYHUDAICLFK-UHFFFAOYSA-N
osmium tetroxide,osmium tetraoxide,osmic acid,perosmic oxide,osmic acid anhydride,osmium viii oxide,oso4,perosmic acid anhydride,osmiumtetroxide,osmium oxide, t-4
30318
tetraoxoosmium
O=[Os](=O)(=O)=O
20816-12-0
O4Os
254.226
MFCD00011150
VUVGYHUDAICLFK-UHFFFAOYSA-N
osmium tetroxide,osmium tetraoxide,osmic acid,perosmic oxide,osmic acid anhydride,osmium viii oxide,oso4,perosmic acid anhydride,osmiumtetroxide,osmium oxide, t-4
30318
tetraoxoosmium
O=[Os](=O)(=O)=O

Reagent Grade, 99.8%

  • ≥95% for general laboratory use.
Linear Formula OsO4
UN Number 2471
Formula Weight 254.23g/mol
Percent Purity 99%8
Grade ReagentPlus™
Vapor Pressure 7mmHg (20°C)
Vapor Density 8.8 (vs air)
Quantity 1g
Chemical Name or Material Osmium Tetroxide

P260-P280-P284-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P342 + P311-P403 + P233

H300 + H310 + H330-H314-H334

EINECSNumber : 244-058-7

RTECSNumber : RN1140000